3,5-Bis(trifluoromethyl)benzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine

PC3104M-10G PC3104M-1G PC3104M-5G
77414-468EA 37.7 USD
77414-468 77414-470 77414-472
3,5-Bis(trifluoromethyl)benzylamine 95%
3,5-Bis(trifluoromethyl)benzylamine
Formula: C₉H₇F₆N
Boiling Pt: 82…84 °C/15 mmHg
Melting Pt: 50…55 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009909
CAS Number: 85068-29-7
EINECS: 285-290-9
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR