2-Chloro-6-fluorobenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1864D-10G PC1864D-25G PC1864D-5G
77413-376EA 30.28 USD
77413-376 77413-378 77413-380
2-Chloro-6-fluorobenzylamine 98%
2-Chloro-6-fluorobenzylamine
Formula: C₇H₇ClFN
Boiling Pt: 91…93 °C/15 mmHg
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042458
CAS Number: 15205-15-9
EINECS: 239-259-1
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR