1,1,3,3-Tetramethyl-2-thiourea 98%

Supplier: APOLLO SCIENTIFIC

OR927378-100G OR927378-25G OR927378-500G
77411-898EA 63.16 USD
77411-898 77411-900 77411-902
1,1,3,3-Tetramethyl-2-thiourea 98%
1,1,3,3-Tetramethyl-2-thiourea
Formula: C₅H₁₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD00008324
CAS Number: 2782-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR