(3,3,3-Trifluoropropyl)trichlorosilane 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trichloro(3,3,3-trifluoropropyl)silane

PC7823-25G PC7823-5G
77418-244EA 152.82 USD
77418-244 77418-246
(3,3,3-Trifluoropropyl)trichlorosilane 97%
(3,3,3-Trifluoropropyl)trichlorosilane
Formula: F₃CCH₂CH₂SiCl₃
MW: 231.5 g/mol
Boiling Pt: 113…114 °C
Density: 1.395 g/cm³ (25 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00013602
CAS Number: 592-09-6
EINECS: 209-744-2
UN: 2985
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR