D-(+)-Proline
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
H-D-Pro-OH, (R)-(+)-Proline, D-Proline
OR0718-25G
OR0718-5G
OR0718-100G
77402-200EA
41.78
USD
77402-200
77402-202
77402-198
D-(+)-Proline
D-(+)-Proline
Chiral intermediate for pharmaceuticals and agrochemicals.
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Melting Pt: 223 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064317 CAS Number: 344-25-2 EINECS: 206-452-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...