Iron(III) acetylacetonate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

OR15745-500G OR15745-100G OR15745-25G
77405-554EA 96.78 USD
77405-554 77405-550 77405-552
Iron(III) acetylacetonate
Iron(III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100
Melting Pt: 180…182 °C
Density: 5.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR