Iron(III) fluoride, anhydrous 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Iron trifluoride, Ferric fluoride

PC3291-100G PC3291-50G
77414-678EA 206.82 USD
77414-678 77414-680
Iron(III) fluoride, anhydrous 99%
Iron(III) fluoride
Formula: F₃Fe
MW: 112.84 g/mol
Melting Pt: 1000
Density: 3.52 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011012
CAS Number: 7783-50-8
EINECS: 232-002-4
UN: 3260
ADR: 8 II
Merck Index: 12,04065

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR