Silver tetrafluoroborate 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Argentous tetrafluoroborate

PC6430-100G PC6430-10G PC6430-50G
77417-376EA 316.86 USD
77417-376 77417-378 77417-380
Silver tetrafluoroborate 99%
Silver tetrafluoroborate

Converts alpha-bromoalkanones to the corresponding fluoro-derivative, Tetrahedron Lett., 1979, 3357.Soluble in many organic solvents.

Formula: AgBF₄
MW: 194.67 g/mol
Melting Pt: 200
Storage Temperature: Ambient
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR