tert-Butylchlorodimethylsilane

Supplier: APOLLO SCIENTIFIC
Danger

OR019066-100G OR019066-1KG OR019066-25G
77401-860EA 24.46 USD
77401-860 77401-862 77401-864
tert-Butylchlorodimethylsilane
tert-Butylchlorodimethylsilane
Formula: C₆H₁₅ClSi
Boiling Pt: 124…126 °C
Melting Pt: 86…89 °C
Density: 0.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4
UN: 2925
ADR: 4.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR