Dibenzoylmethane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Diphenyl-1,3-propanedione, 1,3-Diphenylpropane-1,3-dione

OR18346-100G OR18346-250G
77406-260EA 21.32 USD
77406-260 77406-262
Dibenzoylmethane 98%
Dibenzoylmethane
Formula: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
MW: 224.26 g/mol
Boiling Pt: 219…221 °C/18 mmHg
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00003085
CAS Number: 120-46-7
EINECS: 204-398-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR