4'-Hydroxy-3'-methoxyacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin

OR26691-25G OR26691-5G
77407-824EA 23.4 USD
77407-824 77407-826
4'-Hydroxy-3'-methoxyacetophenone
4'-Hydroxy-3'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 263…265 °C
Melting Pt: 112…115 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008747
CAS Number: 498-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR