ω-Bromo-4-cyanoacetophenone 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone

OR29653-100G OR29653-25G OR29653-5G
77408-414EA 309.72 USD
77408-414 77408-416 77408-418
ω-Bromo-4-cyanoacetophenone 95%
ω-Bromo-4-cyanoacetophenone
Formula: C₉H₆BrNO
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 2923
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR