1,3-Dinitro-4-(trifluoromethoxy)benzene 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
2,4-Dinitro-1-(trifluoromethoxy)benzene, 2,4-Dinitro(trifluoromethoxy)benzene
PC3042-100G
PC3042-25G
77414-416EA
186.44
USD
77414-416
77414-418
1,3-Dinitro-4-(trifluoromethoxy)benzene 98%
1,3-Dinitro-4-(trifluoromethoxy)benzene
Formula:
C₇H₃F₃N₂O₅ MW: 252.11 g/mol Boiling Pt: 273…274 °C Melting Pt: –21 °C Density: 1.623 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042268 CAS Number: 655-07-2 UN: 2927 ADR: 6.1+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...