4-Bromo-2-methoxyphenol 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR22659-1G
OR22659-25G
OR22659-5G
77406-922EA
21.26
USD
77406-922
77406-924
77406-926
4-Bromo-2-methoxyphenol 97%
4-Bromo-2-methoxyphenol
Formula:
C₇H₇BrO₂ Boiling Pt: 129…132 °C/12 mmHg Melting Pt: 34…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051937 CAS Number: 7368-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...