3-(Dimethylamino)-1-propanol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Dimethylamino)propan-1-ol, 3-Dimethylamino-1-propanol

OR5472-100G OR5472-25G OR5472-5G
77410-434EA 303.6 USD
77410-434 77410-436 77410-438
3-(Dimethylamino)-1-propanol 98%
3-(Dimethylamino)-1-propanol
Formula: C₅H₁₃NO
Boiling Pt: 163…164 °C
Melting Pt: –35 °C
Density: 0.88 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002945
CAS Number: 3179-63-3
UN: 2734
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR