Trimethylsilylacetylene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethynyltrimethylsilane

OR18355-100G OR18355-25G OR18355-500G
77406-270EA 65.88 USD
77406-270 77406-272 77406-274
Trimethylsilylacetylene 97%
Trimethylsilylacetylene
Formula: C₅H₁₀Si
MW: 98.22 g/mol
Boiling Pt: 52…53 °C
Density: 0.71 g/cm³ (25 °C)
Flash Pt: –34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008569
CAS Number: 1066-54-2
EINECS: 213-919-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR