Cyclopropanecarbonyl chloride 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR17255-100G
OR17255-250G
OR17255-25G
77405-996EA
63.18
USD
77405-996
77405-998
77406-000
Cyclopropanecarbonyl chloride 98%
Cyclopropanecarbonyl chloride
Formula:
C₄H₅ClO Boiling Pt: 119…121 °C Density: 1.152 g/cm³ (25 °C) Flash Pt: 23 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001277 CAS Number: 4023-34-1 EINECS: 223-684-4 UN: 3286 ADR: 3+6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...