2,2'-Difluorobiphenyl

Supplier: APOLLO SCIENTIFIC
Warning

PC2748-1G PC2748-25G PC2748-5G
77413-828EA 82.42 USD
77413-828 77413-830 77413-832
2,2'-Difluorobiphenyl
2,2'-Difluorobiphenyl
Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
CAS Number: 388-82-9
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR