Pentamethylbenzenesuphonyl chloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR23675-100G OR23675-25G OR23675-5G
77407-362EA 692.14 USD
77407-362 77407-364 77407-366
Pentamethylbenzenesuphonyl chloride, Technical Grade
Pentamethylbenzenesuphonyl chloride
Formula: C₁₁H₁₅ClO₂S
Melting Pt: 81…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014722
CAS Number: 52499-94-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR