3,5-Bis(trifluoromethyl)phenyl isothiocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3115B-100G PC3115B-25G PC3115B-5G
77414-498EA 241.46 USD
77414-498 77414-500 77414-502
3,5-Bis(trifluoromethyl)phenyl isothiocyanate 97%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate
Formula: C₉H₃F₆NS
Boiling Pt: 109…110 °C/35 mmHg
Density: 1.485 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00040838
CAS Number: 23165-29-9
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR