2,3-Dimethylbenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR30107-1G OR30107-25G OR30107-5G
77408-746EA 28.52 USD
77408-746 77408-748 77408-750
2,3-Dimethylbenzoyl chloride
2,3-Dimethylbenzoyl chloride
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 122…124 °C
Melting Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00045217
CAS Number: 21900-46-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR