Toluene-4-sulfonamide 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR18741-100G
OR18741-1KG
OR18741-500G
77406-382EA
21.26
USD
77406-382
77406-384
77406-386
Toluene-4-sulfonamide 98%
Formula:
C₇H₉NO₂S Boiling Pt: 221 °C/10 mmHg Melting Pt: 136…140 °C Flash Pt: 202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011692 CAS Number: 70-55-3 EINECS: 200-741-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...