2,4-Dimethoxybenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

OR28314-100G OR28314-25G OR28314-500G
77408-064EA 111.06 USD
77408-064 77408-066 77408-068
2,4-Dimethoxybenzyl alcohol
2,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 177…179 °C/10 mmHg
Melting Pt: 38…40 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004614
CAS Number: 7314-44-5
EINECS: 230-775-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR