2-Methoxybenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Hydroxymethyl)anisole, (2-Methoxyphenyl)methanol

OR2641-100G OR2641-500G
77407-782EA 151.8 USD
77407-782 77407-786
2-Methoxybenzyl alcohol 98%
2-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 248…250 °C
Density: 1.039 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004611
CAS Number: 612-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR