2-tert-Butyl-4-methoxyphenol 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-BHA, 4-Hydroxy-3-tert-butylanisole, 3-tert-Butyl-4-hydroxyanisole

OR930692-100G OR930692-25G OR930692-500G
77411-942EA 63.16 USD
77411-942 77411-944 77411-946
2-tert-Butyl-4-methoxyphenol 95%
2-tert-Butyl-4-methoxyphenol
Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040484
CAS Number: 121-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR