3-Iodobenzyl alcohol, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-Iodophenyl)methanol, 3-Iodobenzylalcohol

OR16142-100G OR16142-25G OR16142-5G
77405-640EA 289.34 USD
77405-640 77405-642 77405-644
3-Iodobenzyl alcohol, Technical Grade
3-Iodobenzyl alcohol
Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 254…255 °C
Density: 1.842 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004635
CAS Number: 57455-06-8
EINECS: 260-744-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR