α,α,α-Trifluoro-m-toluoyl chloride 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7600-100G
PC7600-25G
PC7600-500G
77418-020EA
49.92
USD
77418-020
77418-022
77418-024
α,α,α-Trifluoro-m-toluoyl chloride 98%
α,α,α-Trifluoro-m-toluoyl chloride
Formula:
C₈H₄ClF₃O Boiling Pt: 184…186 °C Density: 1.38 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000680 CAS Number: 2251-65-2 EINECS: 218-844-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...