2-Fluoro-3-(trifluoromethyl)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4374M-25G PC4374M-5G
77415-292EA 140.6 USD
77415-292 77415-294
2-Fluoro-3-(trifluoromethyl)benzoyl chloride 97%
2-Fluoro-3-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃ClF₄O
Boiling Pt: 192…194 °C
Density: 1.517 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00061154
CAS Number: 208173-19-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR