(2,3-Difluorophenyl)acetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC2874EF-100G PC2874EF-25G PC2874EF-5G
77413-940EA 206.82 USD
77413-940 77413-942 77413-944
(2,3-Difluorophenyl)acetonitrile 97%
(2,3-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00061277
CAS Number: 145689-34-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR