2,4-Dichloro-6-methylphenyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Warning

OR0354-1G OR0354-5G
77402-052EA 81.34 USD
77402-052 77402-054
2,4-Dichloro-6-methylphenyl isothiocyanate
2,4-Dichloro-6-methylphenyl isothiocyanate
Formula: C₈H₅Cl₂NS
Melting Pt: 57…58 °C
Storage Temperature: Ambient
CAS Number: 306935-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR