4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2074NM-1G PC2074NM-5G
77413-508EA 95.76 USD
77413-508 77413-510
4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate 97%
4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Formula: C₈H₃ClF₃NS
MW: 237.63 g/mol
Boiling Pt: 248…251 °C
Melting Pt: 34…36 °C
Density: 1.47 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041090
CAS Number: 23163-86-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR