α-Bromo-3-chloro-2-fluorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-3-chloro-2-fluorobenzene

PC1864G-100G PC1864G-25G PC1864G-5G
77413-388EA 120.22 USD
77413-388 77413-390 77413-392
α-Bromo-3-chloro-2-fluorotoluene 97%
α-Bromo-3-chloro-2-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 125…126 °C/15 mmHg
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00042503
CAS Number: 85070-47-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR