α-Bromo-2-chloro-4-fluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide

PC0070-10G PC0070-25G PC0070-100G
77412-374EA 21.26 USD
77412-374 77412-376 77412-372
α-Bromo-2-chloro-4-fluorotoluene 98%
α-Bromo-2-chloro-4-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47
Boiling Pt: 97…99 °C/10 mmHg
Melting Pt: 33…39 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00236025
CAS Number: 45767-66-6
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR