ω-Bromo-3-chloroacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-3'-chloroacetophenone, Bromomethyl m-chlorophenyl ketone, 2-Bromo-1-(3-chlorophenyl)ethanone, 3-Chlorophenacyl bromide, α-Bromo-3-chloroacetophenone

OR0271-1G OR0271-25G OR0271-5G
77401-984EA 21.26 USD
77401-984 77401-986 77401-988
ω-Bromo-3-chloroacetophenone 97%
ω-Bromo-3-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Melting Pt: 37…40 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792669
CAS Number: 41011-01-2
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR