Ethyl-4-iodobenzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

OR10986-100G OR10986-25G OR10986-500G
77403-012EA 44.64 USD
77403-012 77403-014 77403-016
Ethyl-4-iodobenzoate 98%
Ethyl-4-iodobenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 281…283 °C
Density: 1.65 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017344
CAS Number: 51934-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR