2,6-Difluorobenzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,6-Difluorobenzene-1-sulphonyl chloride

PC2646-100G PC2646-25G
77413-738EA 158.94 USD
77413-738 77413-740
2,6-Difluorobenzenesulfonyl chloride 97%
2,6-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 105…110 °C/2 mmHg
Density: 1.568 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00085004
CAS Number: 60230-36-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR