3-(Trifluoromethoxy)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Trifluoromethoxy)benzene-1-sulphonyl chloride

PC0934-100G PC0934-25G PC0934-5G
77412-746EA 210.9 USD
77412-746 77412-748 77412-750
3-(Trifluoromethoxy)benzenesulfonyl chloride 97%
3-(Trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 229…230 °C
Density: 1.53 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01091016
CAS Number: 220227-84-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR