3-(Trifluoromethoxy)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7438M-1G PC7438M-5G
77417-902EA 28.52 USD
77417-902 77417-904
3-(Trifluoromethoxy)benzoyl chloride 97%
3-(Trifluoromethoxy)benzoyl chloride
Formula: C₈H₄ClF₃O₂
Density: 1.43 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061268
CAS Number: 86270-03-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR