2,5-Dichlorobenzenesulphonyl chloride 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
2,5-Dichlorobenzenesulphonyl Chloride, 2,5-Dichlorobenzene-1-sulphonyl chloride
OR5382-100G
OR5382-25G
77410-334EA
186.44
USD
77410-334
77410-336
2,5-Dichlorobenzenesulphonyl chloride 98%
2,5-Dichlorobenzenesulphonyl chloride
Formula:
C₆H₃Cl₃O₂S MW: 245.51 Melting Pt: 39 Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007429 CAS Number: 5402-73-3 EINECS: 226-453-6 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...