3-Chloro-2-fluorobenzenesulphonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Chloro-2-fluorobenzene-1-sulphonyl chloride

PC4233-100G PC4233-25G PC4233-5G
77415-216EA 152.74 USD
77415-216 77415-218 77415-220
3-Chloro-2-fluorobenzenesulphonyl chloride 97%
3-Chloro-2-fluorobenzenesulphonyl chloride
Formula: C₆H₃Cl₂FO₂S
Density: 1.584 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094234
CAS Number: 351003-48-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR