2-Chloro-6-fluorobenzoyl chloride 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC1863W-100G
PC1863W-25G
PC1863W-5G
77413-366EA
109.02
USD
77413-366
77413-368
77413-370
2-Chloro-6-fluorobenzoyl chloride 97%
2-Chloro-6-fluorobenzoyl chloride
Formula:
C₇H₃Cl₂FO Boiling Pt: 106…108 °C/15 mmHg Density: 1.43 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00178746 CAS Number: 79455-63-3 EINECS: 279-162-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...