4-Bromo-2-fluorophenyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503014-25G PC503014-5G
77416-326EA 247.58 USD
77416-326 77416-328
4-Bromo-2-fluorophenyl isothiocyanate
4-Bromo-2-fluorophenyl isothiocyanate
Formula: C₇H₃BrFNS
Melting Pt: 37…42 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
CAS Number: 81171-71-3
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR