Silver trifluoromethanesulphonate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt
PC6450-100G
PC6450-25G
PC6450-5G
77417-398EA
211.5
USD
77417-398
77417-400
77417-402
Silver trifluoromethanesulphonate 98%
Silver trifluoromethanesulphonate
Increases leaving group ability of halogens; can be used for conversion of alkyl halides to triflates & acyl halides to acyl triflates; soluble in ether and other organic solvents.
Formula:
CAgF₃O₃S MW: 256.94 g/mol Melting Pt: 356 Storage Temperature: Ambient |
MDL Number:
MFCD00013226 CAS Number: 2923-28-6 EINECS: 220-882-2 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...