Ethyl trifluoromethanesulphonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3284-25G PC3284-5G
77414-656EA 189.2 USD
77414-656 77414-658
Ethyl trifluoromethanesulphonate 98%
Ethyl trifluoromethanesulphonate
Formula: C₃H₅F₃O₃S
MW: 178.13 g/mol
Boiling Pt: 115
Density: 1.374 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00000410
CAS Number: 425-75-2
EINECS: 207-037-3
UN: 2920
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR