(S)-(-)-α-Methylbenzylamine 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine

OR28829-100G OR28829-500G
77408-252EA 24.46 USD
77408-252 77408-254
(S)-(-)-α-Methylbenzylamine 99%
(S)-(-)-α-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 187…189 °C
Melting Pt: –10 °C
Density: 0.952 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064406
CAS Number: 2627-86-3
EINECS: 220-098-0
UN: 2922
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR