(R)-(+)-α-Methylbenzylamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine

OR5862-100G OR5862-25G
77410-548EA 37.7 USD
77410-548 77410-550
(R)-(+)-α-Methylbenzylamine
(R)-(+)-α-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 180…186 °C
Melting Pt: –10 °C
Density: 0.952 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR