Yttrium(III) chloride hexahydrate 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Yttrium trichloride hexahydrate

IN3841-100G IN3841-25G
77401-616EA 254.7 USD
77401-616 77401-618
Yttrium(III) chloride hexahydrate 99,9%
Yttrium(III) chloride hexahydrate
Formula: YCl₃·6H₂O
MW: 303.36
Boiling Pt: 1507
Melting Pt: 153 (dec.)
Density: 2.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149941
CAS Number: 10025-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR