Tin(IV) fluoride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6580-1G PC6580-5G
77417-454EA 108 USD
77417-454 77417-456
Tin(IV) fluoride 98%
Tin(IV) fluoride
Formula: SnF₄
Melting Pt: 705 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00042541
CAS Number: 7783-62-2
EINECS: 232-016-0
UN: 2923
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR