Nickel (II) oxide 99.99+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

IN2704-100G IN2704-50G
77401-508EA 268.96 USD
77401-508 77401-510
Nickel (II) oxide 99.99+%
Nickel (II) oxide
Formula: NiO
Melting Pt: 1900 °C
Density: 6.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR