Nickel(II) chloride, anhydrous 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Nickel dichloride

IN2677-250G IN2677-50G
77401-500EA 330.1 USD
77401-500 77401-502
Nickel(II) chloride, anhydrous 98%
Nickel(II) chloride
Formula: Cl₂Ni
Melting Pt: 1001 °C
Density: 3.55 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR